1x1 image pixel for data collection
E-Gel™ 1 Kb Plus Express DNA Ladder
E-Gel™ 1 Kb Plus Express DNA Ladder
Invitrogen™

E-Gel™ 1 Kb Plus Express DNA Ladder

Green features
Invitrogen E-Gel 1 Kb Plus Express DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in theRead more
Have Questions?
Catalog NumberQuantity
10488091100 applications
Catalog number 10488091
Price (USD)
221.00
Special offer
Online exclusive
248.00
Save 27.00 (11%)
Each
In stock
Add to cart
Quantity:
100 applications
Recurring order eligible. Learn more »
Price (USD)
221.00
Special offer
Online exclusive
248.00
Save 27.00 (11%)
Each
Add to cart
Ask our AI about this Product
Invitrogen E-Gel 1 Kb Plus Express DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 5,000 bp. E-Gel 1 Kb Plus Express DNA Ladder consists of nine individual chromatography-purified DNA fragments with reference bands at 1,500 and 500 bp for easy orientation.

E-Gel 1 Kb Plus Express DNA Ladder is specifically formulated for optimal performance on pre-cast E-Gel agarose gels.

Highlights of E-Gel 1 Kb Plus Express DNA Ladder:
Performance—optimized for use on E-Gel CloneWell ll agarose gels
Sharp, clear bands—chromatography-purified fragments for consistent and reliable results, stable at room temperature up to six months
Ready to use—premixed with loading buffer
Convenient—provided with 1X E-Gel Sample Loading Buffer
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 0.8% E-Gel CloneWell II and E-Gel EX agarose gels. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.04 μg/μL
Gel CompatibilityCloneWell™ II E-Gels, Single Comb E-Gels
Green FeaturesSustainable packaging
Product TypeDNA Ladder
Quantity100 applications
Ready to LoadYes
Sample Loading Volume1.25 mL
Shelf Life6 Months
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Volume (Metric)2 x 1.25 mL
Gel TypeAgarose
Size Range100 to 5000 bp
Unit SizeEach
Contents & Storage
• 2 x 1.25 mL E-Gel 1 Kb Plus Express DNA Ladder
• 1.25 mL 1X E-Gel Sample Loading Buffer

Store at room temperature or at 4°C for up to 6 months.
For longer periods store at -20°C.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3270382Certificate of AnalysisJun 27, 202510488091
3253833Certificate of AnalysisJun 04, 202510488091
3253907Certificate of AnalysisJun 04, 202510488091
3204636Certificate of AnalysisApr 04, 202510488091
3167205Certificate of AnalysisFeb 11, 202510488091
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Sequences of Invitrogen DNA and RNA ladders are proprietary.

Invitrogen DNA ladders contain linear dsDNA fragments.

Invitrogen DNA ladders are composed of double-stranded DNA fragments only.

This can happen if the marker was heated. Please ensure that the ladders are not heated before use.

TrackIt DNA Ladders contain two tracking dyes in the sample buffers, which serve as visual markers for tracking electrophoresis progress through the gel, and also to indicate when maximum resolution is achieved. The tracking dyes should not obscure your visualization of DNA bands in the ladder, as the dyes run outside the limits of most DNA bands in the ladder.

TrackIt Cyan/Orange Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Orange G. We recommend TrackIt Cyan/Orange Loading Buffer for DNA fragments between 10 bp and 1 kb.

The TrackIt Cyan/Yellow Loading Buffer is formulated with unique tracking dyes, Xylene Cyanol FF and Tartrazine. We recommend TrackIt Cyan/Yellow Loading Buffer for DNA fragments between 100 bp and 10 kb. The molecular weights are Xylene Cyanol FF, 638.6; Orange G, 452.4; Tartrazine, 534.4.

Note: The TrackIt DNA Ladders are not recommended for use with polyacrylamide gels and are not designed for quantitation.

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection