Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Citric acid, 99%, pure, anhydrous
Thermo Scientific Chemicals

Citric acid, 99%, pure, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 110450050
Price (USD)
256.65
Special offer
Online exclusive
302.00
Save 45.35 (15%)
Each
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
256.65
Special offer
Online exclusive
302.00
Save 45.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0427603Certificate of AnalysisAug 28, 2023110450250, 110450050, 110450010
A0453307Certificate of AnalysisAug 25, 2023110450250, 110450050, 110450010
A0393282Certificate of AnalysisDec 15, 2022110450250, 110450050, 110450010
A0393283Certificate of AnalysisDec 15, 2022110450250, 110450050, 110450010
A0410335Certificate of AnalysisDec 13, 2022110450250, 110450050, 110450010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection