1,3-Diamino-2-hydroxypropane, 95%, Thermo Scientific Chemicals
1,3-Diamino-2-hydroxypropane, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Diamino-2-hydroxypropane, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 112220250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1,3-diaminopropan-2-ol
Molecular FormulaC3H10N2O
InChI KeyUYBWIEGTWASWSR-UHFFFAOYSA-N
SMILESNCC(O)CN
Molecular Weight (g/mol)90.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only