Chemical Identifiers
CAS54-42-2
IUPAC Name1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11IN2O5
InChI KeyXQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
View more