Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum ether, extra pure, boiling range 40-60°C, Thermo Scientific Chemicals

数量:
2.5 L
25 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 124860025
価格(JPY)
-
見積もりを依頼する
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
Color scale=<10 APHA
Density(20°C) 0.64 to 0.66 g/ml
Residue after evaporation=<0.002 %
Appearance (Form)Clear liquid
Boiling point40°C to 60°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

こちらの製品には複数の製品番号が設定されています。