Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Sodium phosphate dibasic dihydrate, 98+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate dibasic dihydrate, 98+%, for analysis

CAS: 10028-24-7 | H5Na2O6P | 177.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 124992500
Price (USD)
37.40
Each
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
37.40
Each
Add to cart
Chemical Identifiers
CAS10028-24-7
IUPAC Namedisodium dihydrate hydrogen phosphate
Molecular FormulaH5Na2O6P
InChI KeyKDQPSPMLNJTZAL-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White
Titration excess HCl98.0 to 102.0 %
Insoluble matter=<0.3 % (in water)
pH8.4 to 9.6 (1% in water)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469374Certificate of AnalysisFeb 06, 2025124992500, 124990010, 124990050
A0462305Certificate of AnalysisMay 07, 2024124992500, 124990010, 124990050
A0458007Certificate of AnalysisDec 05, 2023124992500, 124990010, 124990050
A0451846Certificate of AnalysisMay 02, 2023124992500, 124990010, 124990050
A0400576Certificate of AnalysisDec 15, 2022124992500, 124990010, 124990050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link