Invitrogen™

AcTEV™ Protease

Catalog number: 12575015
Have Questions?
Invitrogen™

AcTEV™ Protease

Catalog number: 12575015
Have Questions?
Catalog Number
12575015
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
125750151,000 units
-
1257502310,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
AcTEV™ Protease specifically recognizes a seven amino acid sequence (Glu-Asn-Leu-Tyr-Phe-Gln-Gly, cleaving between Gln and Gly), making it useful for removing affinity tags from fusion proteins. AcTEV™ Protease is an improved version of Tobacco Etch Virus (TEV) protease that is highly site-specific, highly active, and significantly more stable than native TEV protease, resulting in enhanced long-term activity. AcTEV™ Protease features:

• Highly specific cleavage activity
• Enhanced enzyme stability for prolonged protease activity (see figure)
• Activity over a broad temperature (+4°C to 30°C) and pH (6.0 to 8.5) range
• Six-histidine sequence to facilitate its removal from the digested protein sample
• Greater than 85% single-band purity with no nonspecific protease contamination

Applications
Incubation with AcTEV™ Protease releases the protein of interest from the fusion tag. This is an effective way to remove solubility, secretion, detection, and purification tags from recombinant proteins.

Enzyme specifications
Purified from E. coli expressing the AcTEV™ Protease gene.

Unit definition
One unit of AcTEV™ Protease cleaves 85% of a 3 μg control substrate in 1 hr at 30°C.

Unit reaction conditions
50 mM Tris-HCl (pH 8.0), 0.5 mM EDTA, 1 mM DTT, 3 μg control substrate, and 1 unit enzyme in 30 μL for 1 hr at 30°C. AcTEV™ Protease is functionally tested for the absence of any nonspecific protease activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1,000 Units
Shipping Condition
Dry Ice
Enzyme
TEV Protease
Compatible Buffer
TEV Buffer
Optimal Reaction Temperature
+4°C to +30°C
Quantity
1000 U
Product Type
Protease
Product Line
AcTEV™

Contents & Storage

AcTEV™ Protease is supplied with a vial of 20X TEV buffer [1 M Tris-HCl (pH 8.0), 10 mM EDTA] and a vial of 100 mM DTT.

Store at -20° C. Guaranteed stable for 1 year when properly stored.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text