s-Trioxane, 99+%, Thermo Scientific Chemicals
s-Trioxane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

s-Trioxane, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
2 kg
Packaging:
View all
Plastic drum
Catalog number 140290051
Price (USD)/ Each
191.65
Online exclusive
213.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
191.65
Online exclusive
213.00 
Save 21.35 (10%)
Add to cart
s-Trioxane, 99+%, Thermo Scientific Chemicals
Catalog number140290051
Price (USD)/ Each
191.65
Online exclusive
213.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-88-3
IUPAC Name1,3,5-trioxane
Molecular FormulaC3H6O3
InChI KeyBGJSXRVXTHVRSN-UHFFFAOYSA-N
SMILESC1OCOCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point60°C to 63°C
Appearance (Color)Colorless to white
Appearance (Form)Crystals or crystalline flakes
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only