Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Catalog Number
146008
Unit Size
Case of 10
Description
Easy Fill Replacement Vent Cap
For Use With (Equipment)
EasyFill Cell Factory Systems
Price (USD)
28.78
Availability
-
Full specifications
Quantity
Description
Easy Fill Replacement Vent Cap
Material
HDPE
For Use With (Equipment)
EasyFill Cell Factory Systems
No. per Case
10
No. per Pack
10
Product Type
System Accessories
Green Features
Fewer resources used and less waste, Sustainable packaging
Plug a media bag or tubing set into any cell factory
Save time and reduce costs
Cleaning processes and sterility validations are minimized
Most accessories are gamma irradiated and individually wrapped (Note: Connector Cat. Nos- 140099 and 173250 are not available gamma irradiated)
Prevent buildup of back pressure during filling application
Venting options provide additional air venting when filling and harvesting by gravity or with a peristaltic pump
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.