Thermo Scientific Chemicals

8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 148451000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 148451000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 148451000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
207386-91-2
IUPAC Name
bis(quinolin-8-ol) sulfuric acid hydrate
Molecular Formula
C18H18N2O7S
InChI Key
BNCXJZDIJIVJJO-UHFFFAOYSA-N
SMILES
O.OS(O)(=O)=O.OC1=C2N=CC=CC2=CC=C1.OC1=C2N=CC=CC2=CC=C1
Appearance (Form)
Powder or crystals
Titration with CHA
97.5 to 102.5 %
Water
=<5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text