Valeric Acid, 99%
Thermo Scientific Chemicals

Valeric Acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 149571000
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
68.60 
Save 9.95 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
68.60 
Save 9.95 (15%)
Add to cart
Valeric Acid, 99%
Catalog number149571000
Price (USD)/ Each
58.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
68.60 
Save 9.95 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepentanoic acid
Molecular FormulaC5H10O2
InChI KeyNQPDZGIKBAWPEJ-UHFFFAOYSA-N
SMILESCCCCC(O)=O
Molecular Weight (g/mol)102.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only