Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip (5 pack) |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Quantity | 25 Plates |
Unit Size | 5 x 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Black, 96-well |
Detection Method | Fluorescence |
Product Line | Pierce |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Black, 96-well |
Detection Method | Fluorescence |
Product Line | Pierce |
Color | Black |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Black, 384-well |
Detection Method | Fluorescence |
Product Line | Pierce |
Color | Black |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 384 |
Surface Treatment | Streptavidin |
Well Shape | Flat Bottom (F-well) |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 8-well Strip |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | Blocker™ BSA Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 96-well (5 pack) |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
Surface Treatment | Streptavidin |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Quantity | 25 Plates |
Unit Size | 5 x 5 plates |
Binding Property | Standard Capacity, Affinity |
Description | Clear, 384-well |
Detection Method | Colorimetric |
Product Line | Pierce |
Color | Clear |
Product Type | Detection Plate, Immunoassay, ELISA |
Material | Polystyrene |
No. of Wells | 384 |
Surface Treatment | Streptavidin |
Well Shape | Flat Bottom (F-well) |
No. per Pack | 5 Plates |
Plate Blocking | SuperBlock™ Buffer |
Unit Size | 5 plates |
Catalog Number | Specifications | Unit Size | Description | Plate Blocking | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
15122 | 5 x 5 plates | Clear, 8-well Strip (5 pack) | SuperBlock™ Buffer | 1,085.65 Online price 1,130.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15219 | 5 plates | Black, 96-well | Blocker™ BSA Buffer | 258.65 Online price 269.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15119 | 5 plates | Black, 96-well | SuperBlock™ Buffer | 266.65 Online price 277.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15407 | 5 plates | Black, 384-well | SuperBlock™ Buffer | 461.65 Online price 480.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15121 | 5 plates | Clear, 8-well Strip | Blocker™ BSA Buffer | 264.65 Online price 272.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15120 | 5 plates | Clear, 8-well Strip | SuperBlock™ Buffer | 261.65 Online price 272.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15125 | 5 plates | Clear, 96-well | Blocker™ BSA Buffer | 227.65 Online price 236.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15124 | 5 plates | Clear, 96-well | SuperBlock™ Buffer | 237.65 Online price 247.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15126 | 5 x 5 plates | Clear, 96-well (5 pack) | SuperBlock™ Buffer | 954.65 Online price 994.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
15405 | 5 plates | Clear, 384-well | SuperBlock™ Buffer | 483.65 Online price 504.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific Pierce Streptavidin Coated Plates are pre-blocked and ready-to-use polystyrene plates for binding biotinylated antibodies or probes for ELISA and other target-specific assays.
Features of Streptavidin Coated Plates include:
• Approximate binding capacity is ∼5 pmol biotin/well
• Easy and gentle immobilization of biotin-containing conjugates
• Low nonspecific binding
• No denaturing of the protein component of a conjugate upon binding
• Ideal for binding small biotinylated hydrophilic molecules (e.g., peptides) that typically exhibit poor binding to polystyrene
• Pre-blocked with your choice of Blocker BSA or SuperBlock Blocking Buffer
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.