UltraPure™ Cesium Chloride, Optical Grade
Product Image
Invitrogen™

UltraPure™ Cesium Chloride, Optical Grade

UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPure™ CesiumRead more
Have Questions?
Catalog NumberQuantity
155070231 kg
Catalog number 15507023
Price (USD)
1,390.65
Online Exclusive
1,502.00
Save 111.35 (7%)
Each
In stock
Add to cart
Quantity:
1 kg
Recurring order eligible. Learn more »
Price (USD)
1,390.65
Online Exclusive
1,502.00
Save 111.35 (7%)
Each
Add to cart
Ask our AI about this Product

UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPure™ Cesium Chloride, Optical Grade can be used to prepare plasmids or to isolate phage, virus particles, or cellular RNA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialCesium Chloride
Product LineUltraPure™
Quantity1 kg
Shipping ConditionRoom Temperature
FormPowder
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1024604ACertificate of AnalysisMay 19, 202515507023
1004017ACertificate of AnalysisOct 18, 202415507023
973615ACertificate of AnalysisOct 06, 202215507023
922448ACertificate of AnalysisOct 20, 202115507023
885080ACertificate of AnalysisJun 26, 202015507023
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection