2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals
2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 159261000
Price (USD)/ Each
84.65
Online exclusive
94.00 
Save 9.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.00 
Save 9.35 (10%)
Add to cart
2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals
Catalog number159261000
Price (USD)/ Each
84.65
Online exclusive
94.00 
Save 9.35 (10%)
-
Add to cart
Chemical Identifiers
CAS507-19-7
IUPAC Name2-bromo-2-methylpropane
Molecular FormulaC4H9Br
InChI KeyRKSOPLXZQNSWAS-UHFFFAOYSA-N
SMILESCC(C)(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Refractive index1.4269 to 1.4289 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only