Chalcone, 97%
Chalcone, 97%
Chalcone, 97%
Thermo Scientific Chemicals

Chalcone, 97%

CAS: 94-41-7 | C15H12O | 208.26 g/mol
数量:
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 159431000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS94-41-7
IUPAC Name(2E)-1,3-diphenylprop-2-en-1-one
Molecular FormulaC15H12O
InChI KeyDQFBYFPFKXHELB-VAWYXSNFSA-N
SMILESO=C(\C=C\C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Melting point53°C to 58°C
Appearance (Color)Yellow
Appearance (Form)Granular powder
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

こちらの製品には複数の製品番号が設定されています。