N-Acetyl-L-cysteine, 98%
N-Acetyl-L-cysteine, 98%
Thermo Scientific Chemicals

N-Acetyl-L-cysteine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 160280500
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
Add to cart
N-Acetyl-L-cysteine, 98%
Catalog number160280500
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS616-91-1
IUPAC Name2-acetamido-3-sulfanylpropanoic acid
Molecular FormulaC5H9NO3S
InChI KeyPWKSKIMOESPYIA-UHFFFAOYNA-N
SMILESCC(=O)NC(CS)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 %
Loss on drying=<0.5 %
Residue after ignition=<0.5 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only