Pierce™ Firefly Luc One-Step Glow Assay Kit
Pierce™ Firefly Luc One-Step Glow Assay Kit
Thermo Scientific™

Pierce™ Firefly Luc One-Step Glow Assay Kit

The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
161971000 Reaction Kit
16196100 Reaction Kit
Catalog number 16197
Price (USD)
788.65
Online Exclusive
890.00
Save 101.35 (11%)
Each
In stock
Add to cart
Quantity:
1000 Reaction Kit
Recurring order eligible. Learn more »
Price (USD)
788.65
Online Exclusive
890.00
Save 101.35 (11%)
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific Pierce Firefly One-Step Luc Glow Assay Kit measures firefly luciferase activity in mammalian cell culture using a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications.

Features of the Firefly One-Step Luc Glow Assay Kit:

Sensitive—highly sensitive detection of firefly luciferase activity
Odorless—reagents have no foul or toxic odor, eliminating the need to work in a fume hood
Convenient—one-step, mix-and-read protocol; no additional reagents required
Stable—increased signal half-life and stability compared to flash assays
Simple—does not require a luminometer with injectors
Automation-friendly—amenable to high throughput assays

This glow-type luciferase assay kit is designed for use with cultured mammalian cells that are transfected with vectors expressing the appropriate firefly luciferase reporter enzyme. Several features make the kit especially suitable for luminometers without injectors and for high-throughput screening (HTS) applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate wells containing the transfected, treated cells. The stable signal output (glow) of this system provides flexibility with regard to incubation time. When used with Thermo Scientific Firefly Luc Plasmids, the kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

Includes:
Assay buffer and substrate solution (100X)

Requires:
Firefly luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Applications:
• Promoter studies for analyzing cis- and trans-acting regulatory elements
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Protein localization reporter assays
• Signal transduction pathway analysis

Firefly luciferase is a 62kDa protein produced in nature by several species of the Lampyridae family of beetles, which includes the genera Photinus and Luciola. Thermo Scientific Red Firefly Luciferase has an emission range of 560-700nm (λmax = 613nm). Bioluminescent signal from firefly luciferase originates from the oxidation of D-Luciferin. Light output captured using a luminometer can be correlated with the amount of firefly luciferase protein produced and used to determine the activity of the promoter driving firefly luciferase expression. Firefly luciferase bioluminescence provides researchers with a non-radioactive assay for measuring transcriptional activity of regulatory elements. Firefly luciferase reporter activity can be used to determine promoter activity, to study upstream pathway activity in the cell, or to determine effect of activators or inhibitors.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayReporter Enzyme, Luciferase Reporter Assay
Compatible CellsMammalian Cells
Detection MethodBioluminescence
For Use With (Equipment)Luminometer (Microplate)
Format384-well plate, 96-well plate
Label TypeEnzyme Labeled
Quantity1000 Reaction Kit
SubstrateD-Luciferin
Substrate PropertiesChemical Substrate
Substrate TypeLuciferase Substrate
Sufficient For1000 Luciferase Assays in Microplate Wells
TargetLuciferase, Firefly Luciferase, Cypridina Luciferase
TechniqueEnhanced Chemiluminescence
TypeAssay Kit
Unit SizeEach
Contents & Storage
• Firefly Luc One-Step Substrate, Lyophilized (makes 1 mL of 1X), store at -20°C
• Firefly Luc One-Step Assay Buffer, 115 mL, store at -20°C

Upon receipt store kit at -20°C.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

I prepared my Luciferase Glow Assay Working Solution four hours ago, can I still use it?

All Luciferase Glow Assay Working Solutions should be stored protected from light.

  • Renilla Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 8 hours at room temperature.
  • Gaussia Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 4 hours at room temperature. 
  • Cypridina Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 2 hours at room temperature.
  • Firefly Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for 4 hours at room temperature. For long term use, store at -20 degrees C for up to 2 months.



  • Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

    What is the benefit of using the TurboLuc Luciferase One-Step Glow Assay Kit over the Luciferase Glow Assay kits?

    TurboLuc Luciferase One-Step Glow Assay Kit measures activity in mammalian cells with a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications. Several features of the TurboLuc Luciferase One-Step Glow Assay Kit make it suitable for luminometers without injectors and for HTS applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate containing the transfected, treated cells. The stable signal output (glow enzyme kinetics) of this system provides flexibility with regard to lead time before read. When used with Thermo Scientific TurboLuc 16 Luciferase vectors, the TurboLuc Luciferase One-Step Glow Assay Kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

    Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

    Figures

    Customers who viewed this item also viewed



    Documents & Downloads

    Certificates

    Lot #Certificate TypeDateCatalog Number(s)
    3213925Certificate of AnalysisJun 11, 202516197
    3214593ACertificate of AnalysisMay 23, 202516196
    3214593Certificate of AnalysisMay 23, 202516196
    AC408971ACertificate of AnalysisApr 10, 202516197
    AC408971Certificate of AnalysisApr 10, 202516197
    5 results displayed, search above for a specific certificate

    Safety Data Sheets

    Other products to consider



    Share catalog number, name or link

    1x1 image pixel for data collection