Thermo Scientific Chemicals

1,4-Dioxane, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167800010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

1,4-Dioxane, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 167800010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 167800010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-91-1
IUPAC Name
1,4-dioxane
Molecular Formula
C4H8O2
InChI Key
RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES
C1COCCO1
Color scale
=<10 APHA
GC
>=99.0 %
Water
=<0.03 %
Appearance (Form)
Clear liquid
UV
at 225 nm A: =<0.5

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text