alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
Thermo Scientific Chemicals

alpha-D(+)-Glucose, 99+%, anhydrous

CAS: 492-62-6 | C6H12O6 | 180.16 g/mol
数量:
1 kg
2.5 kg
25 kg
製品番号(カタログ番号) 170080250
価格(JPY)
-
見積もりを依頼する
数量:
25 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS492-62-6
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotationon anhydrous substance
Appearance (Color)White to almost white
Specific optical rotation+52.5° to +53.3° (20°C, 589 nm) (c=10, H2O/diluteNH3)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。