Thermo Scientific Chemicals

Phosgene iminium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 172211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Phosgene iminium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 172211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 172211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33842-02-3
IUPAC Name
dichloro(dimethylamino)methylium chloride
Molecular Formula
C3H6Cl3N
InChI Key
NRNFKRFWZQQDMD-UHFFFAOYSA-M
SMILES
[Cl-].CN(C)[C+](Cl)Cl
Titration Argentometric
>=97.5 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text