2-Iodo-1,1,1-trifluoroethane, 99%, Thermo Scientific Chemicals
2-Iodo-1,1,1-trifluoroethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Iodo-1,1,1-trifluoroethane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number 176080500
Price (USD)/ Each
302.00
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
302.00
Add to cart
2-Iodo-1,1,1-trifluoroethane, 99%, Thermo Scientific Chemicals
Catalog number176080500
Price (USD)/ Each
302.00
-
Add to cart
Chemical Identifiers
CAS353-83-3
IUPAC Name1,1,1-trifluoro-2-iodoethane
Molecular FormulaC2H2F3I
InChI KeyRKOUFQLNMRAACI-UHFFFAOYSA-N
SMILESFC(F)(F)CI
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4000 to 1.4020 (20°C, 589 nm)
Appearance (Color)Clear colorless to pink
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only