Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Thermo Scientific Chemicals

Methanol, 99.9%, for analysis

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
10 L
25 L
Catalog number 176840025
Price (USD)
69.65
Special offer
Online exclusive
82.00
Save 12.35 (15%)
Each
In stock
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
82.00
Save 12.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 % (K.F.)
Carbonyl compounds=<0.001 %
Residue after evaporation=<0.001 %
KMnO4-reducing substance=<0.00025 %
Calcium (Ca)=<0.2 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2537936Certificate of AnalysisJun 25, 2025176840100, 176840025, 176840250, 176840010, 176845000
2542227Certificate of AnalysisJun 23, 2025176840100, 176840025, 176840250, 176840010, 176845000
2541208Certificate of AnalysisJun 12, 2025176840100, 176840025, 176840250, 176840010, 176845000
2540251Certificate of AnalysisJun 09, 2025176840100, 176840025, 176840250, 176840010, 176845000
2538412Certificate of AnalysisMay 24, 2025176840100, 176840025, 176840250, 176840010, 176845000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection