Thermo Scientific Chemicals

Cellulose acetate, average M.W. 100,000

Catalog number: 177785000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Cellulose acetate, average M.W. 100,000

Catalog number: 177785000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 177785000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-35-7
Appearance (Color)
White
Infrared spectrum
Conforms
Acetyl content
39.8 % (typical)
Hydroxyl content
3.6 % (typical)
Loss on drying
=<5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text