Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

Bacterial Alkaline Phosphatase

Catalog number: 18011015
Have Questions?
Invitrogen™

Bacterial Alkaline Phosphatase

Catalog number: 18011015
Have Questions?
Catalog Number
18011015
Unit Size
2500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
180110152500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Bacterial Alkaline Phosphatase (BAP) removes 3´ and 5´ phosphates from DNA and RNA. BAP is active at 65°C for at least 1 h and is inactivated by phenol extraction.

Applications:
Dephosphorylation of 5´-phosphorylated termini of vector DNA to prevent self-ligation. Dephosphorylation of 5´ termini of nucleic acids prior to forward reaction with kinase.

Source:
Purified from E. coli C90.

Performance and Quality Testing:
Endodeoxyribonuclease, exodeoxyribonuclease, and ribonuclease assays; dephosphorylation
efficiency determined.

Unit Definition:
One unit hydrolyzes 1 nmol of ATP in 30 min. at 37°C.

Unit Reaction Conditions:
10 mM Tris-HCl (pH 8.0), 1.5 mM [γ-32P]ATP, and enzyme in 50 μl for 30 min. at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Wet Ice
Enzyme
BAP
Quantity
2500 U
Product Type
Alkaline Phosphatase

Contents & Storage

Bacterial Alkaline Phosphatase is supplied with a vial of 10X dephosphorylation buffer [100 mM Tris-HCl (pH 8.0)]. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text