Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:For PCR enzyme with ≤0.01 copy of bacterial gDNA per enzyme unit, check out Invitrogen Platinum Taq DNA Polymerase, DNA-free.
Invitrogen™

Taq DNA Polymerase, native

Catalog number: 18038240
Have Questions?
Invitrogen™

Taq DNA Polymerase, native

Catalog number: 18038240
Have Questions?
Catalog Number
18038240
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3 x 500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
180382405,000 units
-
18038018100 units
-
18038042500 units
-
180380673 x 500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Taq DNA Polymerase is a thermostable enzyme that synthesizes DNA from single-stranded templates in the presence of dNTPs and a primer. The enzyme consists of a single polypeptide with a molecular weight of 94 kDa. It has a 5'→3' DNA polymerase activity and a 5'→3' exonuclease activity (see figure).

With Taq DNA Polymerase, you get:

  • Your choice of recombinant or native enzyme
  • Amplification of PCR products up to 5 kb in size
  • An enzyme that is licensed and qualified for PCR

Applications

Amplification of DNA from complex genomic, viral, and plasmid templates, RT-PCR, sequencing ssDNA, and cycle sequencing

Source

Native enzyme is purified from Thermus aquaticus YT1.

Unit definition

One unit of Taq DNA Polymerase is the amount of enzyme required to incorporate 10 nmol of deoxyribonucleotide into DNA in 30 min. at 74°C.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Stand-alone enzyme
GC-Rich PCR Performance
Low
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
Exonuclease Activity
5' - 3'
Product Type
DNA Polymerase
Quantity
5000 Units
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Standard PCR
Concentration
5 U/μL
Fidelity (vs. Taq)
1X
Hot Start
No
No. of Reactions
5000 Reactions
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

Taq DNA Polymerase (5 U/μL), 1000 μL
• 10X PCR buffer (without magnesium), 20 mL
• 50 mM MgCl2, 10 mL

Store at -20°C in a non-frost-free freezer.
Stable for 6 months when stored properly.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text