Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Thermo Scientific Chemicals

Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT

Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL | CAS: 109-99-9 | C4H8O | 72.11 g/mol
数量:
250 mL
2.5 L
25 L
パッケージ:
すべて表示
Metal Drum
製品番号(カタログ番号) 181500250
価格(JPY)
-
見積もりを依頼する
数量:
25 L
パッケージ:
Metal Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
さらに表示
Color scale=<10 APHA
Water=<0.02 % (Coulometric)
Stabilizer75 to 400 ppm BHT
GC>=99.85 %
Peroxides=<0.005 % (as H2O2)
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection