Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™
Thermo Scientific Chemicals

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™

CAS: 12075-68-2 | C6H15Al2Cl3 | 247.51 g/mol
数量:
100 mL
800 mL
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 185461000
価格(JPY)
-
見積もりを依頼する
数量:
100 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12075-68-2
IUPAC Namechlorodiethylalumane; dichloro(ethyl)alumane
Molecular FormulaC6H15Al2Cl3
InChI KeyLDXMUMPEDOQULK-UHFFFAOYSA-K
SMILESCC[Al](Cl)Cl.CC[Al](Cl)CC
さらに表示
Appearance (Form)Solution
Assay14.5 to 15.5 % (on Al basis)
Appearance (Color)Clear colorless
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0453960Certificate of Analysis2025年1月31日185468000, 185461000
A0453961Certificate of Analysis2025年1月31日185468000, 185461000
A0427631Certificate of Analysis2024年2月27日185468000, 185461000
A0427630Certificate of Analysis2024年2月27日185468000, 185461000
A0453960Certificate of Analysis2023年8月25日185468000, 185461000
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection