Appearance (Form)Crystalline powder or crystals or mass and/or
Infrared spectrumConforms
Appearance (Color)White to yellow
Appearance (Form)chunks
Melting point50°C to 56°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.