Poly(propylene glycol), average M.W. 2.000
Poly(propylene glycol), average M.W. 2.000
Poly(propylene glycol), average M.W. 2.000
Poly(propylene glycol), average M.W. 2.000
Thermo Scientific Chemicals

Poly(propylene glycol), average M.W. 2.000

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
数量:
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 192185000
価格(JPY)
-
見積もりを依頼する
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
Color scale=<50 APHA
Refractive index1.4490 to 1.4530 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Acid value=<0.1 mg KOH/g
Viscosity305 to 335 cSt (at 25°C)
さらに表示
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection