Thermo Scientific Chemicals

Aluminum ammonium sulfate dodecahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 192382500
250 g, Each
Thermo Scientific Chemicals

Aluminum ammonium sulfate dodecahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 192382500
250 g, Each
Quantity
Catalog number: 192382500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7784-26-1
IUPAC Name
aluminium(3+) ammonium dodecahydrate disulfate
Molecular Formula
AlH28NO20S2
InChI Key
WZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
Iron (Fe)
=<0.001 %
Sodium (Na)
=<0.01 %
Titration Complexometric
98.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text