Immobilized Trypsin, TPCK Treated (Agarose Resin)
Immobilized Trypsin, TPCK Treated (Agarose Resin)
Thermo Scientific™

Immobilized Trypsin, TPCK Treated (Agarose Resin)

Thermo Scientific Pierce Immobilized TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing,Read more
Have Questions?
Catalog NumberQuantity
202302 mL
Catalog number 20230
Price (USD)
115.65
Online Exclusive
134.00
Save 18.35 (14%)
Each
In stock
Add to cart
Quantity:
2 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
134.00
Save 18.35 (14%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Immobilized TPCK Trypsin is a serine endoprotease that is applicable to amino acid analysis and protein sequencing, mapping and structural studies; the immobilized form allows sample separation after treatment.

Features of Thermo Scientific Pierce Immobilized TPCK Trypsin

• Cleaves at carboxyl side of arginine and lysine residues
• Digestion conditions: pH 7.5-9.0, 37°
• Immobilized for ease of separation from cleavage products or affinity purification of Trypsin Inhibitors

Trypsin has a wide range of applications including amino acid analysis and protein sequencing, mapping and structural studies. Enzymes such as trypsin and chymotrypsin have become important tools in sequencing studies since they are highly selective in their cleavage of peptide bonds. Trypsin cleaves only those peptide bonds in which the carboxyl group is contributed by a lysine or an arginine residue, regardless of the length or amino acid sequence of the chain. Our Immobilized TPCK Trypsin can be substituted for free trypsin in many application and is advantageous because it minimizes autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the trypsin. Immobilized trypsin is also more stable against heat-induced denaturation, resulting in longer maintenance of activity.

Applications:
• Removal of adherent cells from tissue culture flasks
• Preparing tryptic fragments for Edman degradation sequencing
• Immobilized trypsin can be used to purify soybean trypsin inhibitor
• Use our Pierce Trypsin, MS Grade (Part No. 90057) for mass spectrometry workflows

Trypsin is a 23.8kDa pancreatic serine endoprotease derived from trypsinogen, an inactive precursor zymogen, after enzymatic removal of an n-terminal leader sequence by enterokinase. Once some trypsin has been formed it can catalyze the conversion of more trypsinogen into its catalytically-active form. Trypsin is treated with L-1-tosylamido-2-phenylethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotrypsin activity without affecting trypsin activity.

Related Products
Trypsin, TPCK Treated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Immobilized Trypsin, TPCK Treated
FormLiquid Suspension
Quantity2 mL
Product TypeTrypsin
Unit SizeEach
Contents & Storage
Contents: Immobilized TPCK Trypsin, 2 mL of settled gel supplied as a 50% slurry containing glycerol and 0.05% sodium azide.
Storage: Upon receipt store at 4°C.
Product is shipped at ambient temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
AB411508Certificate of AnalysisMar 07, 202520230
AB405409Certificate of AnalysisFeb 12, 202520230
ZI397105Certificate of AnalysisOct 15, 202420230
SPECSCertificate of AnalysisOct 15, 202420230
ZE394930Certificate of AnalysisMay 29, 202420230
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link