1x1 image pixel for data collection
Pierce™ DTT (Dithiothreitol)
Pierce™ DTT (Dithiothreitol)
Thermo Scientific™

Pierce™ DTT (Dithiothreitol)

Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE).Related ProductsBond-Breaker™ TCEP Solution, NeutralRead more
Have Questions?
Catalog NumberQuantity
202905 g
Catalog number 20290
Price (USD)
193.00
Each
In stock
Add to cart
Quantity:
5 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
193.00
Each
Add to cart
Ask our AI about this Product
Dithiothreitol is a popular reducing agent for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE).

Related Products
Bond-Breaker™ TCEP Solution, Neutral pH
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce DTT, Cleland's Reagent
FormPowder
Product LinePierce™
Quantity5 g
Product TypeDithiothreitol
Unit SizeEach
Contents & Storage
Store at 4°C.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How do I prepare a 1 M DTT solution from powder and how should I store the solution?

Preparing 1 M DTT:
1) Dissolve 3.09 g DTT in 20 mL distilled H2O.
2) Dispense into 1 mL aliquots.
3) Store the aliquots at -20 degrees C.
Note: Upon thawing the vial, there may be some precipitation that can be re-dissolved by warming and mixing.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Can you please recommend a method for reduction, alkylation, and digestion of an antibody?

Prepare 1 mg/mL antibody in 7.0 M Guanidine HCl, 100 mM Tris, pH 8.3. For reduction, add DTT (Cat. Nos. A39255, 20290) solution at 10 mM final concentration and incubate for 30 mins at room temperature. For alkylation, add sodium iodoacetate at 20 mM final concentration and incubate in the dark for 20 mins at room temperature. Perform buffer exchange followed by digestion using Pierce MS grade Trypsin, Cat. Nos. 90058, 90057 (1:10 enzyme to substrate ratio).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3213256Certificate of AnalysisMay 21, 202520290
AC387868Certificate of AnalysisApr 08, 202520290
AA387867Certificate of AnalysisFeb 12, 2025DNU-20290, 20290
ZJ386492Certificate of AnalysisDec 16, 202420290
ZH386423Certificate of AnalysisOct 15, 202420290
5 results displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection