1x1 image pixel for data collection
Sodium orthovanadate, 99%
Sodium orthovanadate, 99%
Sodium orthovanadate, 99%
Sodium orthovanadate, 99%
Thermo Scientific Chemicals

Sodium orthovanadate, 99%

CAS: 13721-39-6 | Na3O4V | 183.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 205330500
Price (USD)
135.65
Online Exclusive
151.00
Save 15.35 (10%)
Each
In stock
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
135.65
Online Exclusive
151.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13721-39-6
IUPAC Nametrisodium oxovanadiumtris(olate)
Molecular FormulaNa3O4V
InChI KeyIHIXIJGXTJIKRB-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[O-][V]([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Silicon (Si)=<200 ppm
Appearance (Color)White to off-white
Assay>=98.5 % (as Na3VO4)
Potassium (K)=<150 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473153Certificate of AnalysisMay 29, 2025205330500, 205332500
A0468256Certificate of AnalysisJan 31, 2025205330500, 205332500
A0457380Certificate of AnalysisJun 14, 2024205330500, 205332500
A0450666Certificate of AnalysisMay 02, 2023205330500, 205332500
A0403480Certificate of AnalysisDec 15, 2022205330500, 205332500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection