all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
all-trans-Retinoic acid, 97%
Thermo Scientific Chemicals

all-trans-Retinoic acid, 97%

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 302-79-4 | C20H28O2 | 300.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 207341000
Price (USD)
59.30
Each
In stock
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
59.30
Each
Add to cart
Chemical Identifiers
CAS302-79-4
IUPAC Name(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
Molecular FormulaC20H28O2
InChI KeySHGAZHPCJJPHSC-YCNIQYBTSA-N
SMILESC\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point179°C to 184°C
HPLC>=96.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Stephensen, C.B.; Lietz, G. Vitamin A in resistance to and recovery from infection: relevance to SARS-CoV2. Br J Nutr. 2021, 126(11), 1663-1672.
  2. He, J.; Gu, Y.; Zhang, S. Vitamin A and Breast Cancer Survival: A Systematic Review and Meta-analysis. Clin Breast Cancer. 2018, 18(6), e1389-e1400.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472212Certificate of AnalysisMay 17, 2025207340050, 207340010, 207341000
A0469037Certificate of AnalysisJan 31, 2025207340050, 207340010, 207341000
A0465695Certificate of AnalysisSep 02, 2024207340050, 207340010, 207341000
A0463290Certificate of AnalysisJun 14, 2024207340050, 207340010, 207341000
A0460305Certificate of AnalysisFeb 22, 2024207340050, 207340010, 207341000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection