Thermo Scientific Chemicals

Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 213121000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Mercury(I) chloride, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 213121000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 213121000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10112-91-1
IUPAC Name
dimercuranebis(ylium) dichloride
Molecular Formula
Cl2Hg2
InChI Key
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Hg+][Hg+]
Appearance (Color)
Off-white
Titration Iodimetric
>=99.0 %
Mercuric Chloride
=<0.01 % (HgCl2)
Appearance (Form)
Powder
Residue after reduction
=<0.02 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text