Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips
Thermo Scientific™

Extended Length 200μL ART™ Barrier Specialty Filter Pipettor Tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Unit Size
2160PPK
also known as 2160P
200 μLPack of 768
2160P200 μLCase of 3072
Catalog number 2160PPK
also known as 2160P
Price (USD)
118.15
Online Exclusive
256.60
Save 138.45 (54%)
Pack of 768
In stock
Add to cart
Volume (Metric):
200 μL
Unit Size:
Pack of 768
Recurring order eligible. Learn more »
Price (USD)
118.15
Online Exclusive
256.60
Save 138.45 (54%)
Pack of 768
Add to cart
Ask our AI about this Product
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetting for a variety of applications:

Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ ™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART XLP 200, Filtered, Sterile,
Length (English)3.59 in.
Length (Metric)9.12cm
ModelART XLP 200
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)200 μL
Unit SizePack of 768
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Share catalog number, name or link

1x1 image pixel for data collection