1x1 image pixel for data collection
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
ART 1000 REACH RACKED barrier sterile pipette tips
Thermo Scientific™

ART 1000 REACH RACKED barrier sterile pipette tips

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Unit Size
20791000 μLCase of 3200
2079PK
also known as 2079
1000 μLPack of 800
Catalog number 2079
Price (USD)
752.40
Online Exclusive
836.00
Save 83.60 (10%)
Case of 3200
In stock
Add to cart
Volume (Metric):
1000 μL
Unit Size:
Case of 3200
Recurring order eligible. Learn more »
Price (USD)
752.40
Online Exclusive
836.00
Save 83.60 (10%)
Case of 3200
Add to cart
Ask our AI about this Product
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetting for a variety of applications:

Extended length to prevent contamination by keeping the pipette shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 1000 REACH, Filtered, Sterile, Rack
Length (English)4.66 in.
Length (Metric)11.85cm
ModelART 1000 REACH
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRack
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)1000 μL
Unit SizeCase of 3200
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Share catalog number, name or link

1x1 image pixel for data collection