1x1 image pixel for data collection
Carnauba wax, refined, No. 1, yellow
Carnauba wax, refined, No. 1, yellow
Thermo Scientific Chemicals

Carnauba wax, refined, No. 1, yellow

CAS: 8015-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 222382500
Price (USD)
35.65
Special offer
Online exclusive
42.10
Save 6.45 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
42.10
Save 6.45 (15%)
Each
Add to cart
Chemical Identifiers
CAS8015-86-9
SpecificationsSpecification SheetSpecification Sheet
Acid value2 to 7 mg KOH/g
Melting point80°C to 88°C
Saponification value78 to 95 mg KOH/g
Residue after ignition=<0.25 %
Appearance (Form)Flakes
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469203Certificate of AnalysisJan 31, 2025222380050, 222380010, 222382500
A0465223Certificate of AnalysisAug 19, 2024222380050, 222380010, 222382500
A0452224Certificate of AnalysisMay 02, 2023222380050, 222380010, 222382500
A0394358Certificate of AnalysisDec 16, 2022222380050, 222380010, 222382500
A0394306Certificate of AnalysisDec 16, 2022222380050, 222380010, 222382500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection