Thermo Scientific Chemicals

Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals

Catalog number: 223320025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Hydrobromic acid, for analysis, ca. 48% solution in water, Thermo Scientific Chemicals

Catalog number: 223320025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 223320025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10035-10-6
IUPAC Name
hydrogen bromide
Molecular Formula
BrH
InChI Key
CPELXLSAUQHCOX-UHFFFAOYSA-N
SMILES
Br
Appearance (Color)
Clear colorless to light yellow
Titration with NaOH
47.0 to 49.0 %
Iodide (I)
=<30 ppm
Appearance (Form)
Solution
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text