Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Thermo Scientific Chemicals

Saccharin, sodium salt hydrate, 99+%

CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol
数量:
250 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 223370010
価格(JPY)
-
見積もりを依頼する
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS82385-42-0
IUPAC Namesodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate
Molecular FormulaC7H4NNaO3S
InChI KeyOAZGZMKGRPRHJX-UHFFFAOYSA-M
SMILES[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
さらに表示
Benzoate and salicylatePasses test
Benzoate and salicylatePasses test
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

ロット番号Certificate TypeDateCatalog Number(s)
A0468986Certificate of Analysis2025年1月31日223372500, 223370051, 223370010
A0465093Certificate of Analysis2024年8月15日223372500, 223370051, 223370010
A0460424Certificate of Analysis2024年2月27日223372500, 223370051, 223370010
A0453431Certificate of Analysis2023年8月25日223372500, 223370051, 223370010
A0452723Certificate of Analysis2023年5月03日223372500, 223370051, 223370010
5件の結果が表示されました。 上記から特定の証明書を検索します

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection