1x1 image pixel for data collection
Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set
Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set
Thermo Scientific™

Pierce™ Bovine Serum Albumin Standard Pre-Diluted Set

Thermo Scientific Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls inRead more
Have Questions?
Catalog NumberQuantity
232087 x 3.5 mL
Catalog number 23208
Price (USD)
184.65
Online Exclusive
205.00
Save 20.35 (10%)
Each
In stock
Add to cart
Quantity:
7 x 3.5 mL
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
184.65
Online Exclusive
205.00
Save 20.35 (10%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce BSA Protein Assay Standards are high-quality reference samples for generating accurate standard curves and calibration controls in total protein assays.

Features of BSA Protein Assay Standards:

Convenient—complete set containing seven ready-to-use dilutions
Universal—recognized as the industry standard for protein quantitation in colorimetric protein assays
Pure and stable—supplied in ultrapure 0.9% saline solution with 0.05% sodium azide

These bovine serum albumin (BSA) solutions are protein concentration reference standards for use in BCA, Bradford and other protein assay protocols. BSA is the universally accepted reference protein for total protein quantitation. The set of standards is prepared from a stock solution that is calibrated by direct comparison to purified BSA (Fraction V) from the National Institute of Standards and Technology (NIST).

Applications:
• Protein assay quantitation standard (BCA Protein Assay, Coomassie-Bradford Assay, etc.)
• Protein recovery control for desalting and other column procedures
• General calibration of spectrophotometer UV-lamp (absorbance at 280nm)

Selection of a protein standard is potentially the greatest source of error in any protein assay. The best choice for a standard is a purified, known concentration of the most abundant protein contained in the samples being tested. Often, a highly purified, known concentration of the protein of interest is not available or it is too expensive to use as the standard, or the sample itself is a mixture of many proteins (e.g., cell lysate). In such cases, the best standard is one that will produce a normal (i.e., average) color response curve with the selected protein assay method and is readily available to any researcher. BSA is such a protein, and the Pierce Albumin Standards are the most convenient source of ready-to-use BSA standard.

For greatest accuracy in estimating total protein concentration in unknown samples, it is essential to include a standard curve each time the assay is performed. This is particularly true for the protein assay methods that produce non-linear standard curves. Deciding on the number of standards and replicates used to define the standard curve depends upon the degree of non-linearity in the standard curve and the degree of accuracy required. In general, fewer points are needed to construct a standard curve if the color response is linear. Typically, standard curves are constructed using at least two replicates for each point on the curve.

Related Products
Pierce™ Bovine Serum Albumin Standard Ampules, 2 mg/mL
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayProtein Standards
DescriptionPierce Bovine Serum Albumin Standard Pre-Diluted Set
For Use With (Application)Internal Standard
Product LinePierce™
Product TypeProtein Quantitation Assay
Quantity7 x 3.5 mL
SpecificityNot Target-Specific
Unit SizeEach
Contents & Storage
BSA in 0.9% sodium chloride with sodium azide; one each of the following μg/mL concentrations: 125, 250, 500, 750, 1000, 1500, 2000

Store at 2–8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can you provide the shelf-life for the Pierce Bovine Serum Albumin Standard Pre-Diluted Set?

The Pierce Bovine Serum Albumin Standard Pre-Diluted Set is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3212521Certificate of AnalysisMay 07, 202523208
ZL399902Certificate of AnalysisJan 31, 202523208
ZH396279Certificate of AnalysisOct 15, 202423208
ZF392810Certificate of AnalysisJun 20, 202423208
ZB386450Certificate of AnalysisFeb 28, 202423208
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection