Sodium Bicarbonate 7.5% solution
Sodium Bicarbonate 7.5% solution
Gibco™

Sodium Bicarbonate 7.5% solution

Sodium Bicarbonate is buffer commonly used for maintaining the pH of cell culture medium in the presence of 4–10% carbon詳細を見る
製品番号(カタログ番号)数量
25080094100 mL
製品番号(カタログ番号) 25080094
価格(JPY)
3,500
Each
お問い合わせください ›
数量:
100 mL
Ask our AI about this Product
Sodium Bicarbonate is buffer commonly used for maintaining the pH of cell culture medium in the presence of 4–10% carbon dioxide. In addition to buffering, sodium bicarbonate provides some nutritional benefit, while rarely displaying any cell toxicity. Formulated as 75 g per liter of water, Gibco™ Sodium Bicarbonate, 7.5% Solution is perfect for supplementing dry powder medium during reconstitution. The final Sodium Bicarbonate concentration used in the medium depends on the media formulation and the carbon dioxide concentration used in the incubator.

Product Use
For Research Use Only. Not for use in diagnostic procedures.

Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Bicarbonate Solution is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ Sodium Bicarbonate Solution product made in our Scotland facility (25080-060). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
研究用途にのみご使用ください。診断目的には使用できません。
仕様
細胞タイプMammalian Cells
濃度0.075
数量100 mL
品質保持期間18 Months
出荷条件Room Temperature
形状Liquid
製品タイプSodium Bicarbonate
無菌性Sterile-filtered
pH8.3
Unit SizeEach
組成および保存条件
Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

ドキュメントおよびダウンロード

証明書

Safety Data Sheets

こちらの製品には複数の製品番号が設定されています。

1x1 image pixel for data collection