内容Fluorescent red dyed polystyrene microspheres in water
直径(メートル法)平均0.3 μm
直径(メートル法)公称0.3 μm
染色剤タイプEuropium Chelate
内容Package Insert Sheet containing Certicate of Analysis
パーキングエリアBiotin binding of ∼1000pmol/mg
粒子密度1.06g/cm³
数量100 mL
安定性Product stable for a minimum of 24 months
表面修飾Streptavidin
均一性<5% CV
色White
直径(メートル法)0.3 μm
使用対象(アプリケーション)Immunohistological Studies
材料Polystyrene or Polystyrene with Coploymer Grafted Surface Coated with Streptavidin
製品タイプFluorescent Streptavidin-Coated Particles
Unit SizeEach
組成および保存条件
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.