Sodium hydrosulfide hydrate, pure, flakes
Sodium hydrosulfide hydrate, pure, flakes
Sodium hydrosulfide hydrate, pure, flakes
Sodium hydrosulfide hydrate, pure, flakes
Thermo Scientific Chemicals

Sodium hydrosulfide hydrate, pure, flakes

CAS: 207683-19-0 | HNaS | 56.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 296200250
Price (USD)
27.65
Online Exclusive
30.00
Save 2.35 (8%)
Each
In stock
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
27.65
Online Exclusive
30.00
Save 2.35 (8%)
Each
Add to cart
Chemical Identifiers
CAS207683-19-0
IUPAC Namesodium sulfanide
Molecular FormulaHNaS
InChI KeyHYHCSLBZRBJJCH-UHFFFAOYSA-M
SMILES[Na+].[SH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to green to red to brown
AppearanceColor changes in the material may occur
Appearance (Form)Deliquescent flakes
Titration Iodimetric67.0 to 73.0 % (NaHS)
Sodium sulfide (Na2S)=<3.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473810Certificate of AnalysisJun 25, 2025296200100, 296200010, 296200250, 296200025
A0464444Certificate of AnalysisJul 26, 2024296200100, 296200010, 296200250, 296200025
A0459917Certificate of AnalysisFeb 07, 2024296200100, 296200010, 296200250, 296200025
A0440099Certificate of AnalysisSep 15, 2023296200100, 296200010, 296200250, 296200025
A0454277Certificate of AnalysisAug 25, 2023296200100, 296200010, 296200250, 296200025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection