2-Ethylhexyl bromide, 95%
2-Ethylhexyl bromide, 95%
2-Ethylhexyl bromide, 95%
2-Ethylhexyl bromide, 95%
Thermo Scientific Chemicals

2-Ethylhexyl bromide, 95%

CAS: 18908-66-2 | C8H17Br | 193.13 g/mol
数量:
500 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 305335000
価格(JPY)/ Each
-
見積もりを依頼する
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
IUPAC Name3-(bromomethyl)heptane
Molecular FormulaC8H17Br
InChI KeyNZWIYPLSXWYKLH-UHFFFAOYNA-N
SMILESCCCCC(CC)CBr
Molecular Weight (g/mol)193.13
さらに表示
Appearance (Form)Liquid
Refractive index1.4530 to 1.4560 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

こちらの製品には複数の製品番号が設定されています。