Thermo Scientific Chemicals

Tin(II) fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 308505000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tin(II) fluoride, 99%, Thermo Scientific Chemicals

Catalog number: 308505000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 308505000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-47-3
IUPAC Name
λ²-tin(2+) difluoride
Molecular Formula
F2Sn
InChI Key
ANOBYBYXJXCGBS-UHFFFAOYSA-L
SMILES
[F-].[F-].[Sn++]
Appearance (Form)
Powder or crystals
Appearance (Color)
White to almost white
Titration Iodimetric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text