Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Gallium, 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Gallium, 99.99%, (trace metal basis)

CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number 317990100
Price (USD)
95.65
Special offer
Online exclusive
112.00
Save 16.35 (15%)
Each
-
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
112.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS7440-55-3
IUPAC Namegallium
Molecular FormulaGa
InChI KeyGYHNNYVSQQEPJS-UHFFFAOYSA-N
SMILES[Ga]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white to silver grey
Total trace metal impurities=<0.01 %
Appearance (Form)Metallic solid or liquid
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470990Certificate of AnalysisMay 17, 2025317990100, 317990500
A0469694Certificate of AnalysisFeb 12, 2025317990100, 317990500
A0467004Certificate of AnalysisOct 22, 2024317990100, 317990500
A0465656Certificate of AnalysisAug 26, 2024317990100, 317990500
A0460979Certificate of AnalysisMay 27, 2024317990100, 317990500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection