Nimodipine, Thermo Scientific Chemicals
Nimodipine, Thermo Scientific Chemicals
Nimodipine, Thermo Scientific Chemicals
Nimodipine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nimodipine, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
3292800101 g
Catalog number 329280010
Price (USD)
77.65
Online Exclusive
86.40
Save 8.75 (10%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.40
Save 8.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS66085-59-4
IUPAC Name3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC21H26N2O7
InChI KeyUIAGMCDKSXEBJQ-UHFFFAOYNA-N
SMILESCOCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)Yellow
Melting point124°C to 130°C
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0453092Certificate of AnalysisJan 31, 2025329280010
A0453092Certificate of AnalysisAug 25, 2023329280010
A0449485Certificate of AnalysisJan 23, 2023329280010
A0445742Certificate of AnalysisOct 04, 2022329280010
A0355091Certificate of AnalysisJul 22, 2022329280010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection