1x1 image pixel for data collection
Pierce™ DAB Substrate Kit
Pierce™ DAB Substrate Kit
Thermo Scientific™

Pierce™ DAB Substrate Kit

The Thermo Scientific Pierce DAB Substrate Kit enables chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidineRead more
Have Questions?
Catalog NumberQuantity
34002275 mL
Catalog number 34002
Price (USD)
130.65
Online Exclusive
147.00
Save 16.35 (11%)
Each
In stock
Add to cart
Quantity:
275 mL
Request bulk or custom format
Price (USD)
130.65
Online Exclusive
147.00
Save 16.35 (11%)
Each
Add to cart
The Thermo Scientific Pierce DAB Substrate Kit enables chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidine (DAB) in Western blot and tissue staining methods.

DAB (3,3'-diaminobenzidine tetrahydrochloride; MW = 214.1) reacts with HRP in the presence of peroxide to yield an insoluble brown-colored product at locations where peroxidase-conjugated antibodies are bound to samples. The brown precipitate is insoluble in alcohol and other organic solvents, making it an excellent substrate for immunohistochemical staining that requires the use of traditional counterstains and mounting media. The Pierce DAB Substrate Kit includes a 10X solution of DAB and Stable Peroxide Substrate Buffer. Crystalline DAB (powder) is also available.

Features DAB Substrate:

HRP substrate—for detection of horseradish peroxidase activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
Chromogenic—no special equipment needed for visualization; the DAB reaction produces brown bands or spots at sites of reaction with HRP-conjugated antibodies or probes
Package options—choose powder for custom formulation with buffer and peroxide or a complete kit of preformulated solutions for immediate use

Features of the Pierce DAB Substrate Kit:

Easy-to-use—dilute 10X stock solution to 1X in provided stable peroxide buffer
Safer—preformulated DAB solution minimizes safety concerns with this chemical
Complete—includes convenient solution of stable hydrogen peroxide buffer (Part No. 34062)
Stable—store refrigerated at 4°C for at least one year; freezer storage is not necessary

DAB has been used in a variety of applications, including Western blotting, immunohistochemistry and electron microscopy. Many enhancement methods have been reported for DAB involving different buffer conditions, the addition of metal ions and post-treatment applications (Ref. 1-7; also see the Pierce Metal Enhanced DAB Substrate Kit, Part No. 34065). One disadvantage of DAB compared to other substrates is the need to use the working solution immediately because it begins a reaction process upon the addition of hydrogen peroxide. This results in increased background color of the substrate. However, the brown bands resulting from DAB reactions do not fade as quickly as the color-products of other precipitating HRP substrates.

Related Products
DAB Substrate Powder (3,3'-diaminobenzidine tetrahydrochloride)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
DescriptionPierce DAB Substrate Kit
ReactivityHorseradish Peroxidase
Sufficient For25 Uses at 10 mL Each
Detection MethodColorimetric
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce™
Product TypeSubstrate
Quantity275 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Sufficient For: 25 uses at 10 mL each
• DAB (10X), 25 mL
• Stable Peroxide Buffer, 250 mL

Store kit at 4°C.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection