Nalgene™ Coex Polyethylene Film Tank Liners
Nalgene™ Coex Polyethylene Film Tank Liners
Thermo Scientific™

Nalgene™ Coex Polyethylene Film Tank Liners

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)
343050-00055 gal.
343050-00077.5 gal.
343050-001010 gal.
343050-001515 gal.
343050-003030 gal.
343050-005555 gal.
343050-0100100 gal.
343050-0150150 gal.
343050-0200200 gal.
Catalog number 343050-0005
Price (USD)
694.00
Case of 10
In stock
Add to cart
Capacity (English):
5 gal.
Recurring order eligible. Learn more »
Price (USD)
694.00
Case of 10
Add to cart
Ask our AI about this Product

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic

Warranty:
90 days

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
Capacity (English)5 gal.
DescriptionGamma Irradiated: 5 gal. (19L)
For Use With (Equipment)Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case10
No. per Pack10
Capacity (Metric)19 L
MaterialPolyethylene
TypeTank Liner
Unit SizeCase of 10
nalgene-certificate-of-compliance

Certificates of Compliance

Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Nalgene and Nunc products.
Start your request  

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
7350422010Certificate of AnalysisMay 03, 2021343050-0007
7349849010Certificate of AnalysisMay 03, 2021343050-0055
2 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection